MilliporeSigma
  • Mechanisms of Impaired Neutrophil Migration by MicroRNAs in Myelodysplastic Syndromes.
Mechanisms of Impaired Neutrophil Migration by MicroRNAs in Myelodysplastic Syndromes.
Journal of immunology (Baltimore, Md. : 1950) (2017-01-29)
Meiwan Cao, Yayoi Shikama, Hideo Kimura, Hideyoshi Noji, Kazuhiko Ikeda, Tomoyuki Ono, Kazuei Ogawa, Yasuchika Takeishi, Junko Kimura
ABSTRACT
In myelodysplastic syndromes (MDS), functional defects of neutrophils result in high mortality because of infections; however, the molecular basis remains unclear. We recently found that miR-34a and miR-155 were significantly increased in MDS neutrophils. To clarify the effects of the aberrant microRNA expression on neutrophil functions, we introduced miR-34a, miR-155, or control microRNA into neutrophil-like differentiated HL60 cells. Ectopically introduced miR-34a and miR-155 significantly attenuated migration toward chemoattractants fMLF and IL-8, but enhanced degranulation. To clarify the mechanisms for inhibition of migration, we studied the effects of miR-34a and miR-155 on the migration-regulating Rho family members, Cdc42 and Rac1. The introduced miR-34a and miR-155 decreased the fMLF-induced active form of Cdc42 to 29.0 ± 15.9 and 39.7 ± 4.8% of that in the control cells, respectively, although Cdc42 protein levels were not altered. miR-34a decreased a Cdc42-specific guanine nucleotide exchange factor (GEF), dedicator of cytokinesis (DOCK) 8, whereas miR-155 reduced another Cdc42-specific GEF, FYVE, RhoGEF, and PH domain-containing (FGD) 4. The knockdown of DOCK8 and FGD4 by small interfering RNA suppressed Cdc42 activation and fMLF/IL-8-induced migration. miR-155, but not miR-34a, decreased Rac1 protein, and introduction of Rac1 small interfering RNA attenuated Rac1 activation and migration. Neutrophils from patients showed significant attenuation in migration compared with healthy cells, and protein levels of DOCK8, FGD4, and Rac1 were well correlated with migration toward fMLF (r = 0.642, 0.686, and 0.436, respectively) and IL-8 (r = 0.778, 0.659, and 0.606, respectively). Our results indicated that reduction of DOCK8, FGD4, and Rac1 contributes to impaired neutrophil migration in MDS.
Sigma-Aldrich
Anti-phospho-Akt1/PKBα (Ser473) Antibody, clone SK703, rabbit monoclonal, culture supernatant, clone SK703, Upstate®
Pricing and availability is not currently available.