MilliporeSigma
Identification of a Neuronal Receptor Controlling Anaphylaxis.
Cell reports (2016-01-11)
Katarzyna Rogoz, Bejan Aresh, Fabio Batista Freitag, Hanna Pettersson, Elín Ingibjörg Magnúsdóttir, Linn Larsson Ingwall, Helena Haddadi Andersen, Marina Christina Mikaela Franck, Chetan Nagaraja, Klas Kullander, Malin Charlotta Lagerström
ABSTRACT
Allergic reactions can in severe cases induce a state of circulatory shock referred to as anaphylaxis. Histamine, the primary mediator of this condition, is released from immune cells, and, therefore, anaphylaxis has so far been considered an immune system disorder. However, we here show that the glutamatergic receptor mGluR7, expressed on a subpopulation of both peripheral and spinal cord neurons, controls histamine-induced communication through calcium-dependent autoinhibition with implications for anaphylaxis. Genetic ablation of mGluR7, and thus altered regulation of histamine-sensing neurons, caused an anaphylaxis-like state in mGluR7(-/-) mice, which could be reversed by antagonizing signaling between neurons and mast cells but not by antagonizing a central itch pathway. Our findings demonstrate the vital role of nervous system control by mGluR7 in anaphylaxis and open up possibilities for preventive strategies for this life-threatening condition.
Sigma-Aldrich
Anti-mGluR7 Antibody, Upstate®, from rabbit