MilliporeSigma
  • Preparation, application and testing of permanent antibacterial and antiviral coatings.
Preparation, application and testing of permanent antibacterial and antiviral coatings.
Nature protocols (2007-10-20)
Jayanta Haldar, Alisha K Weight, Alexander M Klibanov
ABSTRACT
Protocols for the synthesis of the microbicidal polycation N,N-dodecyl,methyl-polyethylenimine and coating (painting) of glass slides with this polycation's butanol solution are described. Subsequently detailed are the procedures for validating that the resultant coated slides are essentially 100% lethal to the human bacterial pathogens, Staphylococcus aureus and Escherichia coli, as well as to two common strains of influenza virus. The time required to prepare and apply the cationic polymer and to test its microbicidal efficiency is conservatively estimated to be <4 weeks.
Sigma-Aldrich
Select Yeast Extract, EZMix powder, for use in microbial growth medium
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture