MilliporeSigma
  • CD40 Expression in Fibrocytes Is Induced by TSH: Potential Synergistic Immune Activation.
CD40 Expression in Fibrocytes Is Induced by TSH: Potential Synergistic Immune Activation.
PloS one (2016-09-16)
Tünde Mester, Nupur Raychaudhuri, Erin F Gillespie, Hong Chen, Terry J Smith, Raymond S Douglas
ABSTRACT
Fibrocytes appear to participate in inflammation and tissue remodeling in patients with thyroid-associated ophthalmopathy (TAO). These patients have increased frequencies of circulating TSH receptor (TSHR)- and CD40-positive fibrocytes, suggesting TSHR and CD40 may play roles in proinflammatory cytokine production, which ultimately leads to orbital inflammation and tissue remodeling. To investigate the potential interactions between the TSHR and CD40 signaling pathways and their roles in IL-6 and TNF-α production. CD40 expression on fibrocytes was assessed using flow cytometry; IL-6 and TNF-α protein release using Luminex technology; increased IL-6 and TNF-α mRNA abundance, using real-time PCR; TSH- and CD40 ligand (CD40L)-stimulated Akt phosphorylation in fibrocytes, by western blot analysis; TSHR-CD40 protein-protein interaction, using co-immunoprecipitation, and CD40-TSHR co-localization, using immunocytochemistry. TSH enhances CD40 expression at a pre-translational level in fibrocytes. Production of IL-6 and TNF-α after costimulation with TSH and CD40L was greater than that after TSH or CD40L stimulation alone. TSH and CD40L costimulation also resulted in greater Akt phosphorylation. Akt and nuclear factor (NF)-κB inhibitors significantly reduced cytokine production after TSH and CD40L costimulation. TSHR and CD40L are colocalized on the cell surface and form a complex. TSHR and CD40 in fibrocytes appear to be physically and functionally related. TSH stimulates CD40 production on the fibrocyte surface. Cytokine expression upon simultaneous stimulation of TSHR and CD40 is greater than levels achieved with TSH or CD40L alone. Increased expression of CD40 by TSH is a potential mechanism for this process.
Sigma-Aldrich
Milli-Mark® Anti-Collagen Type I-FITC Antibody, clone 5D8-G9, clone 5D8-G9, Milli-Mark®, from mouse