MilliporeSigma
  • A fast signal-induced activation of Poly(ADP-ribose) polymerase: a novel downstream target of phospholipase c.
A fast signal-induced activation of Poly(ADP-ribose) polymerase: a novel downstream target of phospholipase c.
The Journal of cell biology (2000-07-26)
S Homburg, L Visochek, N Moran, F Dantzer, E Priel, E Asculai, D Schwartz, V Rotter, N Dekel, M Cohen-Armon
ABSTRACT
We present the first evidence for a fast activation of the nuclear protein poly(ADP-ribose) polymerase (PARP) by signals evoked in the cell membrane, constituting a novel mode of signaling to the cell nucleus. PARP, an abundant, highly conserved, chromatin-bound protein found only in eukaryotes, exclusively catalyzes polyADP-ribosylation of DNA-binding proteins, thereby modulating their activity. Activation of PARP, reportedly induced by formation of DNA breaks, is involved in DNA transcription, replication, and repair. Our findings demonstrate an alternative mechanism: a fast activation of PARP, evoked by inositol 1,4,5,-trisphosphate-Ca(2+) mobilization, that does not involve DNA breaks. These findings identify PARP as a novel downstream target of phospholipase C, and unveil a novel fast signal-induced modification of DNA-binding proteins by polyADP-ribosylation.
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas