MilliporeSigma
  • Pharmacological characterization of novel A3 adenosine receptor-selective antagonists.
Pharmacological characterization of novel A3 adenosine receptor-selective antagonists.
Neuropharmacology (1997-11-19)
K A Jacobson, K S Park, J L Jiang, Y C Kim, M E Olah, G L Stiles, X D Ji
ABSTRACT
The effects of putative A3 adenosine receptor antagonists of three diverse chemical classes (the flavonoid MRS 1067, the 6-phenyl-1,4-dihydropyridines MRS 1097 and MRS 1191, and the triazoloquinazoline MRS 1220) were characterized in receptor binding and functional assays. MRS1067, MRS 1191 and MRS 1220 were found to be competitive in saturation binding studies using the agonist radioligand [125I]AB-MECA (N6-(4-amino-3-iodobenzyl)adenosine-5'-N-methyluronamide) at cloned human brain A3 receptors expressed in HEK-293 cells. Antagonism was demonstrated in functional assays consisting of agonist-induced inhibition of adenylate cyclase and the stimulation of binding of [35S]guanosine 5'-O-(3-thiotriphosphate) ([35S]GTP-gamma-S) to the associated G-proteins. MRS 1220 and MRS 1191, with KB values of 1.7 and 92 nM, respectively, proved to be highly selective for human A3 receptor vs human A1 receptor-mediated effects on adenylate cyclase. In addition, MRS 1220 reversed the effect of A3 agonist-elicited inhibition of tumor necrosis factor-alpha formation in the human macrophage U-937 cell line, with an IC50 value of 0.3 microM.
Sigma-Aldrich
MRS 1191, solid
Pricing and availability is not currently available.
Sigma-Aldrich
MRS 1220, solid
Pricing and availability is not currently available.