MilliporeSigma
  • The role of the transient receptor potential (TRP) superfamily of cation-selective channels in the management of the overactive bladder.
The role of the transient receptor potential (TRP) superfamily of cation-selective channels in the management of the overactive bladder.
BJU international (2010-12-16)
Karl-Erik Andersson, Christian Gratzke, Petter Hedlund
ABSTRACT
• The pathophysiology of lower urinary tract symptoms (LUTS), detrusor overactivity (DO), and the overactive bladder (OAB) syndrome is multifactorial and remains poorly understood. • The transient receptor potential (TRP) channel superfamily has been shown to be involved in nociception and mechanosensory transduction in various organ systems, and studies of the LUT have indicated that several TRP channels, including TRPV1, TRPV2, TRPV4, TRPM8, and TRPA1, are expressed in the bladder, and may act as sensors of stretch and/or chemical irritation. • However, the roles of these individual channels for normal LUT function and in LUTS/DO/OAB, have not been established. • TRPV1 is the channel best investigated. It is widely distributed in LUT structures, but despite extensive information on morphology and function in animal models, the role of this channel in normal human bladder function is still controversial. Conversely, its role in the pathophysiology and treatment of particularly neurogenic DO is well established. • TRPV1 is co-expressed with TRPA1, and TRPA1 is known to be present on capsaicin-sensitive primary sensory neurones. Activation of this channel can induce DO in animal models. • TRPV4 is a Ca(2+)-permeable stretch-activated cation channel, involved in stretch-induced ATP release, and TRPV4-deficient mice exhibit abnormal frequencies of voiding and non-voiding contractions in cystometric experiments. • TRPM8 is a cool receptor expressed in the urothelium and suburothelial sensory fibres. It has been implicated in the bladder-cooling reflex and in idiopathic DO. • The occurrence of other members of the TRP superfamily in the LUT has been reported, but information on their effects on LUT functions is scarce. There seem to be several links between activation of different members of the TRP superfamily and LUTS/DO/OAB, and further exploration of the involvement of these channels in LUT function, normally and in dysfunction, may be rewarding.
Sigma-Aldrich
Anti-Vanilloid Receptor Like Protein Antibody, pain, CT, Chemicon®, from rabbit
Pricing and availability is not currently available.