MilliporeSigma
  • Changes in cytosolic cGMP and calcium in airway smooth muscle relaxed by 3-morpholinosydnonimine.
Changes in cytosolic cGMP and calcium in airway smooth muscle relaxed by 3-morpholinosydnonimine.
The American journal of physiology (1994-01-01)
K A Jones, R R Lorenz, D O Warner, Z S Katusić, G C Sieck
ABSTRACT
Nitrovasodilators relax airway smooth muscle by both guanosine 3',5'-cyclic monophosphate (cGMP)-dependent and cGMP-independent mechanisms and by mechanisms that reduce cytosolic calcium concentration ([Ca2+]i). This study was conducted to determine the relative importance of these mechanisms in relaxation of canine tracheal smooth muscle (CTSM) induced by 3-morpholinosydnonimine (SIN-1). We measured 1) the effect of SIN-1 on force, [cGMP]i, and [Ca2+]i, and 2) the ability of methylene blue (MB) to antagonize SIN-1-induced relaxation and cGMP accumulation. The ratio of fura 2 emission fluorescence intensities due to excitation at 340- and 380-nm wavelengths (F340/F380) was used as an index of [Ca2+]i. In strips contracted with 0.3 microM acetylcholine (ACh, n = 8) or 24 mM KCl (n = 8), SIN-1 (1-100 microM) caused a concentration-dependent decrease in force which was correlated with a concentration-dependent increase in [cGMP]i. MB (10 microM) proportionally attenuated both relaxation and cGMP accumulation. In fura 2-loaded strips contracted with 0.3 microM ACh (n = 7) or 30 mM KCl (n = 7), reductions in force induced by SIN-1 (1-100 microM) were accompanied by decreases in F340/F380. These findings suggest that in CTSM contracted with ACh or KCl, SIN-1 causes relaxation which appears to be mediated by cGMP-dependent mechanisms that reduce [Ca2+]i.