MilliporeSigma
  • Pyrimidopteridine N-Oxide Organic Photoredox Catalysts: Characterization, Application and Non-Covalent Interaction in Solid State.
Pyrimidopteridine N-Oxide Organic Photoredox Catalysts: Characterization, Application and Non-Covalent Interaction in Solid State.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-02-07)
Richy Hauptmann, Andranik Petrosyan, Franziska Fennel, Miguel A Argüello Cordero, Annette-E Surkus, Jola Pospech
ABSTRACT
Herein we report the photo- and electrochemical characterization of pyrimidopteridine N-oxide-based heterocycles. The potential of their application as organic photoredox catalysts is showcased in the photomediated contra-thermodynamic E→Z isomerization of cinnamic acid derivatives and oxidative cyclization of 2-phenyl benzoic acid to benzocoumarin using molecular oxygen as a mild oxidant. Furthermore, unprecedented intermolecular non-covalent n-π-hole interactions in solid state are discussed based on crystallographic and theoretical data.