MilliporeSigma
  • Protein kinase Cε-calcineurin cosignaling downstream of toll-like receptor 4 downregulates fibrosis and induces wound healing gene expression in cardiac myofibroblasts.
Protein kinase Cε-calcineurin cosignaling downstream of toll-like receptor 4 downregulates fibrosis and induces wound healing gene expression in cardiac myofibroblasts.
Molecular and cellular biology (2013-12-04)
Rui F D S Mesquita, Margaret A Paul, Aida Valmaseda, Asvi Francois, Rita Jabr, Shahzia Anjum, Michael S Marber, Vishwanie Budhram-Mahadeo, Richard J Heads
ABSTRACT
The pathways which regulate resolution of inflammation and contribute to positive remodeling of the myocardium following injury are poorly understood. Here we show that protein kinase C epsilon (PKCε) cooperates with the phosphatase calcineurin (CN) to potentiate induction of cardioprotective gene expression while suppressing expression of fibrosis markers. This was achieved by detailed analysis of the regulation of cyclooxygenase 2 (COX-2) expression as a marker gene and by using gene expression profiling to identify genes regulated by coexpression of CN-Aα/PKCε in adult rat cardiac myofibroblasts (ARVFs) on a larger scale. GeneChip analysis of CN-Aα/PKCε-coexpressing ARVFs showed that COX-2 provides a signature for wound healing and is associated with downregulation of fibrosis markers, including connective tissue growth factor (CTGF), fibronectin, and collagens Col1a1, Col3a1, Col6a3, Col11a1, Col12a1, and Col14a1, with concomitant upregulation of cardioprotection markers, including COX-2 itself, lipocalin 2 (LCN2), tissue inhibitor of metalloproteinase 1 (TIMP-1), interleukin-6 (IL-6), and inducible nitric oxide synthase (iNOS). In primary rat cardiomyocyte cultures Toll-like receptor 4 (TLR4) agonist- or PKCε/CN-dependent COX-2 induction occurred in coresident fibroblasts and was blocked by selective inhibition of CN or PKC α/ε or elimination of fibroblasts. Furthermore, ectopic expression of PKCε and CN in ARVFs showed that the effects on COX-2 expression are mediated by specific NFAT sites within the COX-2 promoter as confirmed by site-directed mutagenesis and chromatin immunoprecipitation (ChIP). Therefore, PKCε may negatively regulate adverse myocardial remodeling by cooperating with CN to downregulate fibrosis and induce transcription of cardioprotective wound healing genes, including COX-2.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, purified by gel-filtration chromatography