MilliporeSigma
  • Direct RNA sequencing enables m6A detection in endogenous transcript isoforms at base-specific resolution.
Direct RNA sequencing enables m6A detection in endogenous transcript isoforms at base-specific resolution.
RNA (New York, N.Y.) (2019-10-19)
Daniel A Lorenz, Shashank Sathe, Jaclyn M Einstein, Gene W Yeo
ABSTRACT
Direct RNA sequencing holds great promise for the de novo identification of RNA modifications at single-coordinate resolution; however, interpretation of raw sequencing output to discover modified bases remains a challenge. Using Oxford Nanopore's direct RNA sequencing technology, we developed a random forest classifier trained using experimentally detected N6-methyladenosine (m6A) sites within DRACH motifs. Our software MINES (m6A Identification using Nanopore Sequencing) assigned m6A methylation status to more than 13,000 previously unannotated DRACH sites in endogenous HEK293T transcripts and identified more than 40,000 sites with isoform-level resolution in a human mammary epithelial cell line. These sites displayed sensitivity to the m6A writer, METTL3, and eraser, ALKBH5, respectively. MINES (https://github.com/YeoLab/MINES.git) enables m6A annotation at single coordinate-level resolution from direct RNA nanopore sequencing.
Sigma-Aldrich
Anti-α-Actinin Antibody, clone AT6/172, clone AT6/172, Upstate®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human METTL3