MilliporeSigma
Mitochondrial translocation of Nur77 mediates cardiomyocyte apoptosis.
European heart journal (2011-01-14)
Zhaokang Cheng, Mirko Völkers, Shabana Din, Daniele Avitabile, Mohsin Khan, Natalie Gude, Sadia Mohsin, Tao Bo, Silvia Truffa, Roberto Alvarez, Matt Mason, Kimberlee M Fischer, Mathias H Konstandin, Xiao-kun Zhang, Joan Heller Brown, Mark A Sussman
ABSTRACT
The cascade of events leading to compromised mitochondrial integrity in response to stress is mediated by various combinatorial interactions of pro- and anti-apoptotic molecules. Nur77, an immediate early gene that encodes a nuclear orphan receptor, translocates from the nucleus to mitochondria to induce cytochrome c release and apoptosis in cancer cells in response to various pro-apoptotic treatments. However, the role of Nur77 in the cardiac setting is still unclear. The objective of this study is to determine the physiological relevance and pathophysiological importance of Nur77 in cardiomyocytes. Myocardial Nur77 is upregulated following cardiomyopathic injury and, while expressed in the postnatal myocardium, declines in level within weeks after birth. Nur77 is localized predominantly in cardiomyocyte nuclei under normal conditions where it is not apoptotic, but translocates to mitochondria in response to oxidative stress both in vitro and in vivo. Mitochondrial localization of Nur77 induces cytochrome c release and typical morphological features of apoptosis, including chromatin condensation and DNA fragmentation. Knockdown of Nur77 rescued hydrogen peroxide-induced cardiomyocyte apoptosis. Translocation of Nur77 from the nucleus to the mitochondria in cardiomyocytes results in the loss of mitochondrial integrity and subsequent apoptosis in response to ischaemia/reperfusion injury. Our findings identify Nur77 as a novel mediator of cardiomyocyte apoptosis and warrants further investigation of mitochondrial Nur77 translocation as a mechanism to control cell death in the treatment of ischaemic heart diseases.