MilliporeSigma
  • Inhibition of human peripheral blood mononuclear cell proliferative response by glycosphingolipids from metacestodes of Echinococcus multilocularis.
Inhibition of human peripheral blood mononuclear cell proliferative response by glycosphingolipids from metacestodes of Echinococcus multilocularis.
Infection and immunity (1996-09-01)
F Persat, C Vincent, D Schmitt, M Mojon
ABSTRACT
The effect on human peripheral blood mononuclear cells (PBMCs) of neutral glycosphingolipids extracted from metacestodes of the parasite Echinococcus multilocularis was investigated. Neutral glycosphingolipids inhibited [3H]thymidine uptake by human PBMCs upon stimulation by mitogens such as phytohemagglutinin A and pokeweed mitogen or by allogeneic Burkitt B cells. This effect was dose dependent and was related to a decrease in interleukin 2 (IL-2) synthesis, the expression of IL-2 receptors (CD25) being unmodified. Addition of exogenous recombinant IL-2 restored the cell proliferation. Partial inhibition of immunoglobulin G (IgG), IgA, and IgM synthesis was observed in the supernatant of cell culture in association with the inhibitory effect. Identification of active subfractions contained in the neutral glycosphingolipid fraction was also studied in relation to cell viability. The free ceramide fraction had an inhibitory effect, in part related to cell lysis, particularly at high concentration, while the monogalactosylceramides had a paradoxical effect: as an activator at low concentrations and as an inhibitor at high concentrations, with limited cell survival. The immunogenic neutral glycosphingolipids containing at least two carbohydrate residues, all having a structure based on Gal beta 1-->6Gal, were inhibitors of PBMC proliferation and showed good cell survival. These results suggest that parasite neutral glycosphingolipids may play an immunologically relevant role in alveolar hydatid disease.
Sigma-Aldrich
Galactocerebrosides from bovine brain, ≥97% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Non-hydroxy fatty acid ceramide from bovine brain, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Palmitic acid, ≥99%
Pricing and availability is not currently available.