MilliporeSigma
  • Identification of mycobacterial alpha-glucan as a novel ligand for DC-SIGN: involvement of mycobacterial capsular polysaccharides in host immune modulation.
Identification of mycobacterial alpha-glucan as a novel ligand for DC-SIGN: involvement of mycobacterial capsular polysaccharides in host immune modulation.
Journal of immunology (Baltimore, Md. : 1950) (2009-09-29)
Jeroen Geurtsen, Sunita Chedammi, Joram Mesters, Marlène Cot, Nicole N Driessen, Tounkang Sambou, Ryo Kakutani, Roy Ummels, Janneke Maaskant, Hiroki Takata, Otto Baba, Tatsuo Terashima, Nicolai Bovin, Christina M J E Vandenbroucke-Grauls, Jérôme Nigou, Germain Puzo, Anne Lemassu, Mamadou Daffé, Ben J Appelmelk
ABSTRACT
Mycobacterium tuberculosis possesses a variety of immunomodulatory factors that influence the host immune response. When the bacillus encounters its target cell, the outermost components of its cell envelope are the first to interact. Mycobacteria, including M. tuberculosis, are surrounded by a loosely attached capsule that is mainly composed of proteins and polysaccharides. Although the chemical composition of the capsule is relatively well studied, its biological function is only poorly understood. The aim of this study was to further assess the functional role of the mycobacterial capsule by identifying host receptors that recognize its constituents. We focused on alpha-glucan, which is the dominant capsular polysaccharide. Here we demonstrate that M. tuberculosis alpha-glucan is a novel ligand for the C-type lectin DC-SIGN (dendritic cell-specific ICAM-3-grabbing nonintegrin). By using related glycogen structures, we show that recognition of alpha-glucans by DC-SIGN is a general feature and that the interaction is mediated by internal glucosyl residues. As for mannose-capped lipoarabinomannan, an abundant mycobacterial cell wall-associated glycolipid, binding of alpha-glucan to DC-SIGN stimulated the production of immunosuppressive IL-10 by LPS-activated monocyte-derived dendritic cells. By using specific inhibitors, we show that this IL-10 induction was DC-SIGN-dependent and also required acetylation of NF-kappaB. Finally, we demonstrate that purified M. tuberculosis alpha-glucan, in contrast to what has been reported for fungal alpha-glucan, was unable to activate TLR2.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype abortus equi, purified by phenol extraction