MilliporeSigma
  • Polymer Encapsulants for Threshold Voltage Control in Carbon Nanotube Transistors.
Polymer Encapsulants for Threshold Voltage Control in Carbon Nanotube Transistors.
ACS applied materials & interfaces (2019-09-19)
François Lapointe, Ashish Sapkota, Jianfu Ding, Jacques Lefebvre
ABSTRACT
Although carbon nanotube transistors present outstanding performances based on key metrics, large-scale uniformity and repeatability required in printable electronics depend greatly on proper control of the electrostatic environment. Through a survey of polymer dielectric encapsulants compatible with printing processes, a simple correlation is found between the measured interfacial charge density and the onset of conduction in a transistor, providing a rational route to control the electrical characteristics of carbon nanotube transistors. Smooth and continuous balancing of the properties between unipolar p-type and n-type transport is achieved using a molar fraction series of poly(styrene-co-2-vinylpyridine) statistical copolymers combined with an electron-donating molecule. We further demonstrate the easy fabrication of a p-n diode which shows a modest rectification of 8:1.
Sigma-Aldrich
4-(2,3-Dihydro-1,3-dimethyl-1H-benzimidazol-2-yl)-N,N-dimethylbenzenamine, 97% (HPLC)
Pricing and availability is not currently available.