MilliporeSigma
  • Supramolecular photochemistry of encapsulated caged ortho-nitrobenzyl triggers.
Supramolecular photochemistry of encapsulated caged ortho-nitrobenzyl triggers.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2019-07-28)
Nareshbabu Kamatham, A Mohan Raj, Richard S Givens, José P Da Silva, V Ramamurthy
ABSTRACT
ortho-Nitrobenzyl (oNB) triggers have been extensively used to release various molecules of interest. However, the toxicity and reactivity of the spent chromophore, o-nitrosobenzaldehyde, remains an unaddressed difficulty. In this study we have applied the well-established supramolecular photochemical concepts to retain the spent trigger o-nitrosobenzaldehyde within the organic capsule after release of water-soluble acids and alcohols. The sequestering power of organic capsules for spent chromophores during photorelease from ortho-nitrobenzyl esters, ethers and alcohols is demonstrated with several examples.
Sigma-Aldrich
3,3-Dimethylacrylic acid, 97%