MilliporeSigma
  • Heme oxygenase-1 arrests Leydig cells functions and impairs their regulation by histamine.
Heme oxygenase-1 arrests Leydig cells functions and impairs their regulation by histamine.
Journal of molecular endocrinology (2019-08-16)
Trinidad Raices, María Luisa Varela, Casandra Margarita Monzón, María Florencia Correa Torrado, Romina María Pagotto, Marcos Besio Moreno, Carolina Mondillo, Omar Pedro Pignataro, Elba Nora Pereyra
ABSTRACT
Testicular Leydig cells (LC) are modulated by several pathways, one of them being the histaminergic system. Heme oxygenase-1 (HO-1), whose upregulation comprises the primary response to oxidative noxae, has a central homeostatic role and might dysregulate LC functions when induced. In this report, we aimed to determine how hemin, an HO-1 inducer, affects LC proliferative capacity and whether HO-1 effects on LC functions are reversible. It was also evaluated if HO-1 interacts in any way with histamine, affecting its regulatory action over LC. MA-10 and R2C cell lines and immature rat LC were used as models. Firstly, we show that after a 24-h incubation with 25 µmol/L hemin, LC proliferation is reversibly impaired by cell cycle arrest in G2/M phase, with no evidence of apoptosis induction. Even though steroid production is abrogated after a 48-h exposure to 25 µmol/L hemin, steroidogenesis can be restored to control levels in a time-dependent manner if the inducer is removed from the medium. Regarding HO-1 and histamine interaction, it is shown that hemin abrogates histamine biphasic effect on steroidogenesis and proliferation. Working with histamine receptors agonists, we elucidated that HO-1 induction affects the regulation mediated by receptor types 1, 2 and 4. In summary, HO-1 induction arrests LC functions, inhibiting steroid production and cell cycle progression. Despite their reversibility, HO-1 actions might negatively influence critical phases of LC development and differentiation affecting their function as well as other androgen-dependent organs. What's more, we have described a hitherto unknown interaction between HO-1 induction and histamine effects.
Sigma-Aldrich
2-((3-Trifluoromethyl)phenyl)histamine dimaleate, ≥98% (HPLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade