MilliporeSigma
  • The NLRP3 inflammasome is involved with the pathogenesis of Mayaro virus.
The NLRP3 inflammasome is involved with the pathogenesis of Mayaro virus.
PLoS pathogens (2019-09-04)
Luiza A de Castro-Jorge, Renan V H de Carvalho, Taline M Klein, Carlos H Hiroki, Alexandre H Lopes, Rafaela M Guimarães, Marcílio Jorge Fumagalli, Vitor G Floriano, Mayara R Agostinho, Renata Dezengrini Slhessarenko, Fernando Silva Ramalho, Thiago M Cunha, Fernando Q Cunha, Benedito A L da Fonseca, Dario S Zamboni
ABSTRACT
Mayaro virus (MAYV) is an arbovirus that circulates in Latin America and is emerging as a potential threat to public health. Infected individuals develop Mayaro fever, a severe inflammatory disease characterized by high fever, rash, arthralgia, myalgia and headache. The disease is often associated with a prolonged arthralgia mediated by a chronic inflammation that can last months. Although the immune response against other arboviruses, such as chikungunya virus (CHIKV), dengue virus (DENV) and Zika virus (ZIKV), has been extensively studied, little is known about the pathogenesis of MAYV infection. In this study, we established models of MAYV infection in macrophages and in mice and found that MAYV can replicate in bone marrow-derived macrophages and robustly induce expression of inflammasome proteins, such as NLRP3, ASC, AIM2, and Caspase-1 (CASP1). Infection performed in macrophages derived from Nlrp3-/-, Aim2-/-, Asc-/-and Casp1/11-/-mice indicate that the NLRP3, but not AIM2 inflammasome is essential for production of inflammatory cytokines, such as IL-1β. We also determined that MAYV triggers NLRP3 inflammasome activation by inducing reactive oxygen species (ROS) and potassium efflux. In vivo infections performed in inflammasome-deficient mice indicate that NLRP3 is involved with footpad swelling, inflammation and pain, establishing a role of the NLRP3 inflammasome in the MAYV pathogenesis. Accordingly, we detected higher levels of caspase1-p20, IL-1β and IL-18 in the serum of MAYV-infected patients as compared to healthy individuals, supporting the participation of the NLRP3-inflammasome during MAYV infection in humans.
Sigma-Aldrich
Anti-Interleukin-1β antibody produced in goat, IgG fraction of antiserum