MilliporeSigma
  • Bacteriocin enterocin CRL35 is a modular peptide that induces non-bilayer states in bacterial model membranes.
Bacteriocin enterocin CRL35 is a modular peptide that induces non-bilayer states in bacterial model membranes.
Biochimica et biophysica acta. Biomembranes (2019-11-19)
Carolina Medina Amado, Carlos J Minahk, Eduardo Cilli, Rafael G Oliveira, Fernando G Dupuy
ABSTRACT
The mechanism of action of the anti-Listeria peptide enterocin CRL35 was studied with biophysical tools by using lipid mixtures that mimicked Gram-positive plasma membranes. Langmuir monolayers and infrared spectroscopy indicated that the peptide readily interacted with phospholipid assembled in monolayers and bilayers to produce a dual effect, depending on the acyl chains. Indeed, short chain mixtures were disordered by enterocin CRL35, but the gel-phases of membranes composed by longer acyl chains were clearly stabilized by the bacteriocin. Structural and functional studies indicated that non-bilayer states were formed when liposomes were co-incubated with enterocin CRL35, whereas significant permeabilization could be detected when bilayer and non-bilayer states co-existed. Results can be explained by a two-step model in which the N-terminal of the peptide firstly docks enterocin CRL35 on the lipid surface by means of electrostatic interactions; then, C-terminal triggers membrane perturbation by insertion of hydrophobic α-helix.