MilliporeSigma
  • Chemerin induced by Treponema pallidum predicted membrane protein Tp0965 mediates the activation of endothelial cell via MAPK signaling pathway.
Chemerin induced by Treponema pallidum predicted membrane protein Tp0965 mediates the activation of endothelial cell via MAPK signaling pathway.
Journal of cellular biochemistry (2019-07-20)
Rui-Li Zhang, Qian-Qiu Wang, Li-Jia Yang
ABSTRACT
Chemerin, a chemoattractant protein, is involved in endothelial dysfunction and vascular inflammation in pathological conditions. In a recent study, we observed the upregulation of chemerin in endothelial cells following in vitro treatment with Treponema pallidum. Here, we investigated the role of chemerin in endothelial cells activation induced by the T. pallidum predicted membrane protein Tp0965. Following stimulation of human umbilical vein endothelial cells (HUVECs) with Tp0965, chemerin and its receptor chemerin receptor 23 (ChemR23) were upregulated, companied with elevated expression of Toll-like receptor 2. Furthermore, chemerin from HUVECs activated endothelial cells via chemerin/ChemR23 signaling in an autocrine/paracrine manner, characterized by upregulated expression of intercellular adhesion molecule 1, E-selectin, and matrix metalloproteinase-2. Activation of endothelial cells depended on the mitogen-activated protein kinase signaling pathway. In addition, Tp0965-induced chemerin promoted THP-1-derived macrophages migration to endothelial cells, also via the chemerin/ChemR23 pathway. The RhoA/ROCK signaling pathway was also involved in THP-1-derived macrophages migration in response to chemerin/ChemR23. Our results highlight the role of Tp0965-induced chemerin in endothelial cells dysfunction, which contributes to the immunopathogenesis of vascular inflammation of syphilis.