MilliporeSigma
  • UHRF1 suppresses retrotransposons and cooperates with PRMT5 and PIWI proteins in male germ cells.
UHRF1 suppresses retrotransposons and cooperates with PRMT5 and PIWI proteins in male germ cells.
Nature communications (2019-10-19)
Juan Dong, Xiaoli Wang, Congcong Cao, Yujiao Wen, Akihiko Sakashita, Si Chen, Jin Zhang, Yue Zhang, Liquan Zhou, Mengcheng Luo, Mingxi Liu, Aihua Liao, Satoshi H Namekawa, Shuiqiao Yuan
ABSTRACT
DNA methylation, repressive histone marks, and PIWI-interacting RNA (piRNA) are essential for the control of retrotransposon silencing in the mammalian germline. However, it remains unknown how these repressive epigenetic pathways crosstalk to ensure retrotransposon silencing in the male germline. Here, we show that UHRF1 is responsible for retrotransposon silencing and cooperates with repressive epigenetic pathways in male germ cells. Conditional loss of UHRF1 in postnatal germ cells causes DNA hypomethylation, upregulation of retrotransposons, the activation of a DNA damage response, and switches in the global chromatin status, leading to complete male sterility. Furthermore, we show that UHRF1 interacts with PRMT5, an arginine methyltransferase, to regulate the repressive histone arginine modifications (H4R3me2s and H3R2me2s), and cooperates with the PIWI pathway during spermatogenesis. Collectively, UHRF1 regulates retrotransposon silencing in male germ cells and provides a molecular link between DNA methylation, histone modification, and the PIWI pathway in the germline.
Sigma-Aldrich
Magnesium acetate solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Bouin′s solution, histological fixative