MilliporeSigma
  • Resolution of ångström-scale protein conformational changes by analyzing fluorescence anisotropy.
Resolution of ångström-scale protein conformational changes by analyzing fluorescence anisotropy.
Nature structural & molecular biology (2019-09-07)
John H Lewis, Zhe Lu
ABSTRACT
Conformational changes within typical protein molecules are rapid and small, making their quantitative resolution challenging. These changes generally involve rotational motions and may thus be resolved by determining changes in the orientation of a fluorescent label that assumes a unique orientation in each conformation. Here, by analyzing fluorescence intensities collected using a polarization microscope at a rate of 50 frames per second, we follow the changes of 10-16° in the orientation of a single bifunctional rhodamine molecule attached to a regulator of conductance to K+ (RCK) domain of the MthK channel, and thus, the transitions between its three conformational states, with effective standard deviation (σ) of 2-5°. Based on available crystal structures, the position of the fluorophore's center differs by 3.4-8.1 Å among the states. Thus, the present approach allows the resolution of protein conformational changes involving ångström-scale displacements.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Supelco
Sodium 2-mercaptoethanesulfonate, analytical standard, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium 2-mercaptoethanesulfonate, BioXtra, ≥98.0% (RT)
Pricing and availability is not currently available.