MilliporeSigma
  • Structural Transformations in the Thermal Dehydration of [Cu2(bpa)(btec)(H2O)4]n Coordination Polymer.
Structural Transformations in the Thermal Dehydration of [Cu2(bpa)(btec)(H2O)4]n Coordination Polymer.
Molecules (Basel, Switzerland) (2019-05-16)
Laura Bravo-García, Edurne S Larrea, Beñat Artetxe, Luis Lezama, Juan M Gutiérrez-Zorrilla, María I Arriortua
ABSTRACT
Reactions between pyridinic ligands such as 1,2-bis(4-pyridyl)ethane (bpa) and transition metal cations are a very widespread technique to produce extended coordination polymers such as Metal-Organic Frameworks. In combination with a second ligand these systems could present different topologies and behaviors. In this context, the use of 1,2,4,5-benzenetetracarboxylic acid (H4btec) gave us a novel 2D compound, [Cu2(bpa)(btec)(H2O)4]n (1), which was prepared by microwave-assisted synthesis and structurally characterized by means of single crystal X-ray diffraction. Its thermal behavior was analyzed through thermogravimetric analysis and variable temperature powder X-ray diffraction, concluding that thermal stability is influenced by the coordination water molecules, allowing two sequential thermochromic phase transformations to take place. These transformations were monitored by electronic paramagnetic resonance spectroscopy and magnetic susceptibility measurements. In addition, the crystal structure of the anhydrous compound [Cu2(bpa)(btec)]n (1.ah) was determined. Finally, a topological study was carried out for the bpa ligand considering all the structures deposited in the Cambridge Structural Databased. More than 1000 structures were analyzed and classified into 17 different topologies, according to the role of the ligand.