MilliporeSigma
  • Triptolide attenuates lipopolysaccharide-induced inflammatory responses in human endothelial cells: involvement of NF-κB pathway.
Triptolide attenuates lipopolysaccharide-induced inflammatory responses in human endothelial cells: involvement of NF-κB pathway.
BMC complementary and alternative medicine (2019-08-04)
Chundong Song, Youping Wang, Lin Cui, Fengna Yan, Si Shen
ABSTRACT
Endothelial cell inflammation is a central event in the pathogenesis of numerous cardiovascular diseases, including sepsis and atherosclerosis. Triptolide, a principal bioactive ingredient of Traditional Chinese Medicine Tripterygium wilfordii Hook.F., displays anti-inflammatory actions in vivo. However, the mechanisms underlying these beneficial effects remain undetermined. The present study investigated the effects and possible mechanisms of triptolide on lipopolysaccharide (LPS)-induced inflammatory responses in human umbilical vein endothelial cells (HUVECs). The effects of triptolide on the LPS-induced production and expression of inflammatory molecules, monocyte adhesion and activation of nuclear factor (NF)-κB pathway were examined in cultured HUVECs. In cultured HUVECs, pre-treatment with triptolide dose-dependently attenuated LPS-induced cytokine and chemokine production, adhesion molecule expression and monocyte adhesion. Mechanistically, triptolide was found to dose-dependently inhibit the LPS-induced increases in the DNA binding activity of NF-κB p65 associated with attenuating IκBα phosphorylation and its degradation. Additionally, the present study revealed that triptolide inhibited LPS-triggered NF-κB transcriptional activation in a dose-dependent manner. The results of the present study indicated that triptolide suppresses the inflammatory response of endothelial cells possibly via inhibition of NF-κB activation.