MilliporeSigma
  • Fish pituitary show an active immune response after in vitro stimulation with Vibrio bacterin.
Fish pituitary show an active immune response after in vitro stimulation with Vibrio bacterin.
General and comparative endocrinology (2019-02-16)
Xiaohong Liu, Ali R Khansari, Lluis Tort
ABSTRACT
The pituitary is a central organ of the neuro-endocrine system in fish that plays critical roles in various physiological processes, including stress response and behavior. Although it is known that pituitary hormones can have a direct or indirect influence stimulating or suppressing the immune responses, whether there is a local immune response in the pituitary or what is the effect of the immune stimulus on the pituitary function in fish is unknown. With the aim to understand the interaction between the immune responses and the endocrine axes at the pituitary level, particularly the Hypothalamus-Pituitary-Interrenal (HPI) axis, pituitaries of rainbow trout (Oncorhynchus mykiss) were cultured in vitro, incubated with bacterin, or bacterin plus CRH, cortisol, human recombinant IL1β, or spleen medium for 3 h, and then genes involved in pro-inflammation (il1β, il8, tnfα1, ifnγ), anti-inflammation (tgfβ1b, il10), immune modulation (mhcIIa, c3, mif) and stress response (crhbp, pomca, pomcb, gr1) were tested. Data showed that, incubation with bacterin alone and bacterin plus recombinant IL1β or CRH, as well as medium from bacterin treated spleen caused significant up-regulation of pro-inflammatory genes il1β and il8, while down-regulated the anti-inflammatory gene tgfβ1b. Besides, recombinant IL1β plus bacterin or alone caused raise of mhcIIa and tnfa, respectively. On the contrary, just a slight or even no alteration was recorded in the expression of stress response genes including crhbp, pomca, pomcb and gr1 in the in vitro cultured trout pituitary following this stimulation. These results suggest a local immune gene equipment in the pituitary of fish, and the potential for fish pituitary to develop both innate and adaptive immune responses, whereas that immune stimulation was not able to evoke a significant endocrine stress response in vitro.