MilliporeSigma
  • The hnRNP raly regulates PRMT1 expression and interacts with the ALS-linked protein FUS: implication for reciprocal cellular localization.
The hnRNP raly regulates PRMT1 expression and interacts with the ALS-linked protein FUS: implication for reciprocal cellular localization.
Molecular biology of the cell (2018-10-26)
Lisa Gasperini, Annalisa Rossi, Nicola Cornella, Daniele Peroni, Paola Zuccotti, Valentina Potrich, Alessandro Quattrone, Paolo Macchi
ABSTRACT
RALY is a member of the heterogeneous nuclear ribonucleoprotein family whose transcriptome and interactome have been recently characterized. RALY binds poly-U rich elements within several RNAs and regulates the expression as well as the stability of specific transcripts. Here we show that RALY binds PRMT1 mRNA and regulates its expression. PRMT1 catalyzes the arginine methylation of FUS, an RNA-binding protein that interacts with RALY. We demonstrate that RALY downregulation decreases PRMT1 levels, thus reducing FUS methylation. It is known that mutations in the FUS nuclear localization signal (NLS) retain the protein to the cytosol, promote aggregate formation and are associated with amyotrophic lateral sclerosis. Confirming that inhibiting FUS methylation increases its nuclear import, we report that RALY knockout enhances FUS NLS mutants' nuclear translocation, hence decreasing aggregate formation. Furthermore, we characterize the RNA-dependent interaction of RALY with FUS in motor neurons. We show that mutations in FUS NLS as well as in RALY NLS reciprocally alter their localization and interaction with target mRNAs. These data indicate that RALY's activity is impaired in FUS pathology models, raising the possibility that RALY might modulate disease onset and/or progression.
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture