MilliporeSigma
  • Design and development of multitarget-directed N-Benzylpiperidine analogs as potential candidates for the treatment of Alzheimer's disease.
Design and development of multitarget-directed N-Benzylpiperidine analogs as potential candidates for the treatment of Alzheimer's disease.
European journal of medicinal chemistry (2019-02-21)
Piyoosh Sharma, Avanish Tripathi, Prabhash Nath Tripathi, Santosh Kumar Prajapati, Ankit Seth, Manish Kumar Tripathi, Pavan Srivastava, Vinod Tiwari, Sairam Krishnamurthy, Sushant Kumar Shrivastava
ABSTRACT
The multitarget-directed strategy offers an effective and promising paradigm to treat the complex neurodegenerative disorder, such as Alzheimer's disease (AD). Herein, a series of N-benzylpiperidine analogs (17-31 and 32-46) were designed and synthesized as multi-functional inhibitors of acetylcholinesterase (AChE) and β-secretase-1 (BACE-1) with moderate to excellent inhibitory activities. Among the tested inhibitors, 25, 26, 40, and 41 presented the most significant and balanced inhibition against both the targets. Compounds 40 and 41 exhibited high brain permeability in the PAMPA-BBB assay, significant displacement of propidium iodide from the peripheral anionic site (PAS) of AChE, and were devoid of neurotoxicity towards SH-SY5Y neuroblastoma cell lines up to the maximum tested concentration of 80 μM. Meanwhile, both these compounds inhibited self- and AChE-induced Aβ aggregation in thioflavin T assay, which was also re-affirmed by morphological characterization of Aβ aggregates using atomic force microscopy (AFM). Moreover, 40 and 41 ameliorated the scopolamine-induced cognitive impairment in elevated plus and Y-maze experiments. Ex vivo and biochemical analysis established the brain AChE inhibitory potential and antioxidant properties of these compounds. Further, improvement in Aβ1-42-induced cognitive impairment was also observed by compound 41 in the Morris water maze experiment with significant oral absorption characteristics ascertained by the pharmacokinetic studies.