MilliporeSigma
  • Living annulative π-extension polymerization for graphene nanoribbon synthesis.
Living annulative π-extension polymerization for graphene nanoribbon synthesis.
Nature (2019-06-28)
Yuuta Yano, Nobuhiko Mitoma, Kaho Matsushima, Feijiu Wang, Keisuke Matsui, Akira Takakura, Yuhei Miyauchi, Hideto Ito, Kenichiro Itami
ABSTRACT
The properties of graphene nanoribbons (GNRs)1-5-such as conductivity or semiconductivity, charge mobility and on/off ratio-depend greatly on their width, length and edge structure. Existing bottom-up methods used to synthesize GNRs cannot achieve control over all three of these parameters simultaneously, and length control is particularly challenging because of the nature of step-growth polymerization6-18. Here we describe a living annulative π-extension (APEX)19 polymerization technique that enables rapid and modular synthesis of GNRs, as well as control over their width, edge structure and length. In the presence of palladium/silver salts, o-chloranil and an initiator (phenanthrene or diphenylacetylene), the benzonaphthosilole monomer polymerizes in an annulative manner to furnish fjord-type GNRs. The length of these GNRs can be controlled by simply changing the initiator-to-monomer ratio, achieving the synthesis of GNR block copolymers. This method represents a type of direct C-H arylation polymerization20 and ladder polymerization21, activating two C-H bonds of polycyclic aromatic hydrocarbons and constructing one fused aromatic ring per chain propagation step.