MilliporeSigma
  • Synthesis and Viscosimetric Behavior of Poly(acrylamide-co-2-acrylamido-2-methylpropanesulfonate) Obtained by Conventional and Adiabatic Gel Process via RAFT/MADIX Polymerization.
Synthesis and Viscosimetric Behavior of Poly(acrylamide-co-2-acrylamido-2-methylpropanesulfonate) Obtained by Conventional and Adiabatic Gel Process via RAFT/MADIX Polymerization.
ACS omega (2019-08-29)
Aurora Mejia, Laurent Rodriguez, Charlène Schmitt, Nathalie Andreu, Cedrick Favéro, Olivier Braun, Guillaume Dupuis, Elise Deniau, Stéphanie Reynaud, Bruno Grassl
ABSTRACT
High molar masses homopolymers of both acrylamide (AM) and 2-acrylamido-2-methylpropanesulfonate (AMPS) as well as poly(AM-stat-AMPS) exhibiting a large range copolymer composition has been obtained via the optimization of a purely adiabatic gel process. Monomer concentrations ranging from 2.0 to 3.47 M have been successfully tested while keeping the control of the molar masses up to 5 × 106 g mol-1. The products have been characterized in terms of molecular mass and viscosimetric properties.