MilliporeSigma
  • Improvement of Mechanical Properties and Self-Healing Efficiency by Ex-Situ Incorporation of TiO2 Nanoparticles to a Waterborne Poly(Urethane-Urea).
Improvement of Mechanical Properties and Self-Healing Efficiency by Ex-Situ Incorporation of TiO2 Nanoparticles to a Waterborne Poly(Urethane-Urea).
Polymers (2019-07-25)
Iñigo Díez-García, Arantxa Eceiza, Agnieszka Tercjak
ABSTRACT
This research work was focused on the incorporation of TiO2 nanoparticles into synthesized solvent-free waterborne poly(urethane-urea) (WPUU) based on hydrophilic poly(ethylene oxide) (PU0) in order to improve both the mechanical properties and self-healing effectiveness of a polymer matrix. The incorporation of TiO2 nanoparticles resulted in a successful enhancement of the mechanical properties of nanocomposite films when compared to PU0. Simultaneously, the obtained nanocomposite films did not only maintain the self-healing ability of the PU0 film, measured by means of mechanical properties after successive cutting/recovery cycles, but they also showed a higher self-healing efficiency than the PU0 film. Moreover, the well-dispersed TiO2 nanoparticles, visualized by atomic force microscopy (AFM), kept their conductive properties when embedded in the PU0 matrix, as was confirmed by electrostatic force microscopy (EFM). This research work described a simple and industrially appealing way to control the dispersion of commercially available TiO2 nanoparticles in waterborne poly(urethane-urea) for the designing of inorganic/organic hybrid nanocomposites with enhanced mechanical properties and self-healing efficiency, in which TiO2 nanoparticles preserved their conductive properties within the polymer matrix.