MilliporeSigma
Form of calf diet and the rumen. I: Impact on growth and development.
Journal of dairy science (2019-07-08)
T T Yohe, H Schramm, C L M Parsons, H L M Tucker, B D Enger, N R Hardy, K M Daniels
ABSTRACT
Preweaning diet is known to affect rumen tissue appearance at the gross level. The objectives of this experiment were to investigate effects of different preweaning diets on the growth and development of the rumen epithelium and on putative rumen epithelial stem and progenitor cell measurements at the gene and cell levels. Neonatal Holstein bull calves (n = 11) were individually housed and randomly assigned to 1 of 2 diets. The diets were milk replacer only (MRO; n = 5) or milk replacer with starter (MRS; n = 6). Diets were isoenergetic (3.87 ± 0.06 Mcal of metabolizable energy per day) and isonitrogenous (0.17 ± 0.003 kg/d of apparent digestible protein). Milk replacer was 22% crude protein, 21.5% fat (dry matter basis). The textured calf starter was 21.5% crude protein (dry matter basis). Water was available ad libitum and feed and water intake were recorded daily. Putative stem and progenitor cells were labeled by administering a thymidine analog (5-bromo-2'-deoxyuridine, BrdU; 5 mg/kg of body weight in sterile saline) for 5 consecutive days and allowed a 25-d washout period. Calves were killed at 43 ± 1 d after a 6 h exposure to a defined concentration of volatile fatty acids. We obtained rumen tissue from the ventral sac and used it for immunohistochemical analyses of BrdU (putative stem and progenitor cells) and Ki67 (cell proliferation), gene expression analysis, and morphological measurements via hematoxylin and eosin staining. Epithelial stem and progenitor cell gene markers of interest, analyzed by real-time quantitative PCR, were β1-integrin, keratin-14, notch-1, tumor protein p63, and leucine-rich repeat-containing G protein-coupled receptor 5. Body growth did not differ by diet, but empty reticulorumens were heavier in MRS calves (MRS: 0.67 ± 0.04 kg; MRO: 0.39 ± 0.04 kg). The percentage of label-retaining BrdU basale cells was higher in MRO calves than in MRS calves (2.0 ± 0.3% vs. 0.3 ± 0.2%, respectively). We observed a higher percentage of basale cells undergoing proliferation in MRS calves than in MRO calves (18.4 ± 2.6% vs. 10.8 ± 2.8%, respectively). Rumen epithelial gene expression was not affected by diet, but the submucosa was thicker in MRO calves and the epithelium and corneum/keratin layers were thicker in MRS calves. Presumptive stem and progenitor cells in the rumen epithelium were identifiable by their ability to retain labeled DNA in the long term, changed proliferative status in response to diet, and likely contributed to observed treatment differences in rumen tissue thickness.