- Death-associated protein kinase 3 deficiency alleviates vascular calcification via AMPK-mediated inhibition of endoplasmic reticulum stress.
Death-associated protein kinase 3 deficiency alleviates vascular calcification via AMPK-mediated inhibition of endoplasmic reticulum stress.
Vascular calcification (VC) is a critical feature of chronic kidney disease (CKD), diabetes, hypertension, and atherosclerosis. Death-associated protein kinase 3 (DAPK3) is involved in vascular remodeling in hypertension. However, it remains to be clarified whether DAPK3 controls vascular smooth muscle cell (VSMC) phenotypic transition into an osteogenic cell phenotype, which is an important process for VC. In vivo VC was induced in rats by vitamin D3 and nicotine. VSMCs were incubated with calcifying media containing β-glycerophosphate and Ca2+ to induce VC in vitro. Herein, we demonstrated increased expression of DAPK3 in the aortas of VC rats and VSMCs cultured in calcifying media. Knockdown of DAPK3 significantly inhibited calcifying media-induced VSMC mineralization and retarded the phenotypic transformation of VSMCs into osteogenic cells. Silencing of DAPK3 suppressed endoplasmic reticulum stress (ERS) related protein expressions, but upregulated the phosphorylation level of AMP-activated protein kinase (AMPK) in calcified VSMCs. Moreover, pretreatment with AMPK inhibitor Compound C abolished DAPK3 shRNA-mediated inhibition of ERS in VSMCs. In vivo, DAPK inhibitor significantly prevented calcium deposition in the aortas of VC rats. The present results revealed that DAPK3 modulated VSMC calcification through AMPK-mediated ERS signaling.