MilliporeSigma
  • A Sample Preparation Technique Using Biocompatible Composites for Biomedical Applications.
A Sample Preparation Technique Using Biocompatible Composites for Biomedical Applications.
Molecules (Basel, Switzerland) (2019-04-17)
Huifang Liu, Geun Su Noh, Yange Luan, Zhen Qiao, Bonhan Koo, Yoon Ok Jang, Yong Shin
ABSTRACT
Infectious diseases, especially pathogenic infections, are a growing threat to public health worldwide. Since pathogenic bacteria usually exist in complex matrices at very low concentrations, the development of technology for rapid, convenient, and biocompatible sample enrichment is essential for sensitive diagnostics. In this study, a cucurbit[6]uril (CB) supermolecular decorated amine-functionalized diatom (DA) composite was fabricated to support efficient sample enrichment and in situ nucleic acid preparation from enriched pathogens and cells. CB was introduced to enhance the rate and effectiveness of pathogen absorption using the CB-DA composite. This novel CB-DA composite achieved a capture efficiency of approximately 90% at an Escherichia coli concentration of 10⁶ CFU/mL within 3 min. Real-time PCR analyses of DNA samples recovered using the CB-DA enrichment system showed a four-fold increase in the early amplification signal strength, and this effective method for capturing nucleic acid might be useful for preparing samples for diagnostic systems.