MilliporeSigma
  • Prognostic Role of Circulating Exosomal miR-425-3p for the Response of NSCLC to Platinum-Based Chemotherapy.
Prognostic Role of Circulating Exosomal miR-425-3p for the Response of NSCLC to Platinum-Based Chemotherapy.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2018-09-20)
Daolu Yuwen, Yuzhu Ma, Deqiang Wang, Jian Gao, Xin Li, Wenwen Xue, Minmin Fan, Qiang Xu, Yan Shen, Yongqian Shu
ABSTRACT
Platinum-based doublets with a third-generation agent are the recommended option for many patients with non-small cell lung cancer (NSCLC) with no contraindications to platinum compounds. Unfortunately, the clinical effectiveness of such chemotherapy is limited by intrinsic or acquired resistance. Circulating exosomal miRNAs were isolated and used to perform HiSeq deep-sequencing analyses on serum pool samples from platinum-resistant or platinum-sensitive patients, and six exosomal miRNAs were further validated for their predictive utility by qRT-PCR in 170 serum samples of patients with advanced NSCLC. Gain- and loss-of-function experiments clarified the responsiveness regulating role of the clinically relevant miRNA. IHC analyses were performed to evaluate the association between basal autophagy in lung cancer tissues and responsiveness in 203 patients with NSCLC receiving platinum-based chemotherapy. Six circulating exosomal miRNAs (miR-425-3p, miR-1273h, miR-4755-5p, miR-9-5p, miR-146a-5p, and miR-215-5p) were found to be differentially expressed with the largest fold change in platinum-resistant patients compared with platinum-sensitive patients. High miR-425-3p proved to be a potent predictive biomarker for low responsiveness and poor progression-free survival (PFS). Mechanistically, miR-425-3p upregulated the autophagic levels via targeting AKT1, leading to the decrease in therapeutic response. Concordantly, high levels of basal autophagy in lung cancer tissues correlate with low responsiveness in patients with NSCLC within the early and advanced disease stages. Our study highlights circulating exosomal miR-425-3p as a potential biomarker for improved predictions of the clinical response to platinum-based chemotherapy in patients with NSCLC. This study provides the first evidence that miR-425-3p in NSCLC patient-derived exosomes can be a marker for predicating the clinical response to platinum-based chemotherapy.