MilliporeSigma
  • Presenilin 1 Regulates NF-κB Activation via Association with Breakpoint Cluster Region and Casein Kinase II.
Presenilin 1 Regulates NF-κB Activation via Association with Breakpoint Cluster Region and Casein Kinase II.
Journal of immunology (Baltimore, Md. : 1950) (2018-09-12)
Yuki Tanaka, Lavannya Sabharwal, Mitsutoshi Ota, Ikuma Nakagawa, Jing-Jing Jiang, Yasunobu Arima, Hideki Ogura, Masayasu Okochi, Masaru Ishii, Daisuke Kamimura, Masaaki Murakami
ABSTRACT
We recently reported that NF-κB-mediated inflammation caused by breakpoint cluster region (BCR) is dependent on the α subunit of casein kinase II (CK2α) complex. In the current study, we demonstrate that presenilin 1 (Psen1), which is a catalytic component of the γ-secretase complex and the mutations of which are known to cause familial Alzheimer disease, acts as a scaffold of the BCR-CK2α-p65 complex to induce NF-κB activation. Indeed, Psen1 deficiency in mouse endothelial cells showed a significant reduction of NF-κB p65 recruitment to target gene promoters. Conversely, Psen1 overexpression enhanced reporter activation under NF-κB responsive elements and IL-6 promoter. Furthermore, the transcription of NF-κB target genes was not inhibited by a γ-secretase inhibitor, suggesting that Psen1 regulates NF-κB activation in a manner independent of γ-secretase activity. Mechanistically, Psen1 associated with the BCR-CK2α complex, which is required for phosphorylation of p65 at serine 529. Consistently, TNF-α-induced phosphorylation of p65 at serine 529 was significantly decreased in Psen1-deficient cells. The association of the BCR-CK2α-p65 complex was perturbed in the absence of Psen1. These results suggest that Psen1 functions as a scaffold of the BCR-CK2α-p65 complex and that this signaling cascade could be a novel therapeutic target for various chronic inflammation conditions, including those in Alzheimer disease.