MilliporeSigma
  • Individual variations in platelet reactivity towards ADP, epinephrine, collagen and nitric oxide, and the association to arterial function in young, healthy adults.
Individual variations in platelet reactivity towards ADP, epinephrine, collagen and nitric oxide, and the association to arterial function in young, healthy adults.
Thrombosis research (2018-12-14)
Madelene Lindkvist, Ulrika Fernberg, Liza U Ljungberg, Knut Fälker, Maria Fernström, Anita Hurtig-Wennlöf, Magnus Grenegård
ABSTRACT
Platelet aggregation and secretion can be induced by a large number of endogenous activators, such as collagen, adenosine diphosphate (ADP) and epinephrine. Conversely, the blood vessel endothelium constitutively release platelet inhibitors including nitric oxide (NO) and prostacyclin. NO and prostacyclin are also well-known vasodilators and contribute to alterations in local blood flow and systemic blood pressure. In this study we investigated individual variations in platelet reactivity and arterial functions including blood pressure and flow-mediated vasodilation (FMD) in 43 young, healthy individuals participating in the Lifestyle, Biomarkers and Atherosclerosis (LBA) study. Platelet aggregation and dense granule secretion were measured simultaneously by light transmission and luminescence. FMD was measured with ultrasound. The platelet function assay showed inter-individual differences in platelet reactivity. Specifically, a sub-group of individuals had platelets with an increased response to low concentrations of ADP and epinephrine, but not collagen. When the NO-donor S-nitroso-N-acetyl-DL-penicillamine (SNAP) was combined with high doses of these platelet activators, the results indicated for sub-groups of NO-sensitive and NO-insensitive platelets. The individuals with NO-sensitive platelets in response to SNAP in combination with collagen had a higher capacity of FMD of the arteria brachialis. Platelet reactivity towards ADP, epinephrine and NO differs between young, healthy individuals. Some individuals have a more effective response towards NO, both in the aspect of platelet inhibition ex vivo, as well as vasodilation in vivo.