MilliporeSigma
  • Protective Action of Linear Polyethylenimine against Staphylococcus aureus Colonization and Exaggerated Inflammation in Vitro and in Vivo.
Protective Action of Linear Polyethylenimine against Staphylococcus aureus Colonization and Exaggerated Inflammation in Vitro and in Vivo.
ACS infectious diseases (2019-05-18)
Venkatesh Mayandi, Sreepathy Sridhar, Mobashar Hussain Urf Turabe Fazil, Eunice Tze Leng Goh, Hla Myint Htoon, Gorka Orive, Yeu Khai Choong, Rathi Saravanan, Roger W Beuerman, Timothy Mark Sebastian Barkham, Liang Yang, Mani Baskaran, Vishal Jhanji, Xian Jun Loh, Navin Kumar Verma, Rajamani Lakshminarayanan
ABSTRACT
Increased evolution of multidrug resistant pathogens necessitates the development of multifunctional antimicrobials. There is a perceived need for developing new antimicrobials that can interfere with acute inflammation after bacterial infections. Here, we investigated the therapeutic potential of linear polyethylenimine (LPEI) in vitro and in vivo. The minimum inhibitory concentration of LPEI ranged from 8 to 32 μg/mL and elicited rapid bactericidal activity against clinical isolates of meticillin-resistant Staphylococcus aureus (MRSA). The polymer was biocompatible for human cultured ocular and dermal cells. Prophylactic addition of LPEI inhibited the bacterial colonization of human primary dermal fibroblasts (hDFs). In a scratch wound cell migration assay, LPEI attenuated the migration inhibitory effects of bacterial secretions. The polymer neutralized the cytokine release by hDFs exposed to bacterial secretions, possibly by blocking their accessibility to host cell receptors. Topical instillation of LPEI (1 mg/mL) was noncytotoxic and did not affect the re-epithelialization of injured porcine cornea. In a prophylactic in vivo model of S. aureus keratitis, LPEI was superior to gatifloxacin in terms of reducing stimulation of cytokines, corneal edema, and overall severity of the infection. These observations demonstrate therapeutic potential of LPEI for antimicrobial prophylaxis.