MilliporeSigma
  • N-acetyl-l-cysteine exacerbates kidney dysfunction caused by a chronic high-sodium diet in renal ischemia and reperfusion rats.
N-acetyl-l-cysteine exacerbates kidney dysfunction caused by a chronic high-sodium diet in renal ischemia and reperfusion rats.
Life sciences (2019-06-11)
Carolina Martinez Romão, Rafael Canavel Pereira, Maria Heloisa Massola Shimizu, Luzia Naôko Shinohara Furukawa
ABSTRACT
To investigate the effect of long-term N-acetyl-l-cysteine (NAC) treatment in Wistar rats subjected to renal ischemia and reperfusion (IR) and a chronic high‑sodium diet (HSD). Adult male Wistar rats received an HSD (8.0% NaCl) or a normal‑sodium diet (NSD; 1.3% NaCl) and NAC (600 mg/L) or normal drinking water starting at 8 weeks of age. At 11 weeks of age, the rats from both diet and NAC or water treatment groups underwent renal IR or Sham surgery and were followed for 10 weeks. The study consisted of six animal groups: NSD + Sham + water; NSD + IR + water; NSD + IR + NAC; HSD + Sham + water; HSD + IR + water; and HSD + IR + NAC. Tail blood pressure (tBP) increased with IR and NAC treatment in the NSD group but not in the HSD group. The serum creatinine level was higher after NAC treatment in both diet groups, and creatinine clearance was decreased in only the HSD + IR + NAC group. Albuminuria increased in the HSD + IR + water group and decreased in the HSD + IR + NAC group. Kidney mass was increased in the HSD + IR group and decreased with NAC treatment. Renal fibrosis was prevented with NAC treatment and cardiac fibrosis was decreased with NAC treatment in the HSD + IR group. NAC treatment promoted structural improvements, such as decreased albuminuria and fibrosis, in the kidney and heart. However, NAC could not recover kidney function or blood pressure from the effects of IR associated with an HSD. Therefore, in general, long-term NAC treatment is not effective and is deleterious to recovery of function after kidney injury.