MilliporeSigma
  • Long-term ex vivo haematopoietic-stem-cell expansion allows nonconditioned transplantation.
Long-term ex vivo haematopoietic-stem-cell expansion allows nonconditioned transplantation.
Nature (2019-05-31)
Adam C Wilkinson, Reiko Ishida, Misako Kikuchi, Kazuhiro Sudo, Maiko Morita, Ralph Valentine Crisostomo, Ryo Yamamoto, Kyle M Loh, Yukio Nakamura, Motoo Watanabe, Hiromitsu Nakauchi, Satoshi Yamazaki
ABSTRACT
Multipotent self-renewing haematopoietic stem cells (HSCs) regenerate the adult blood system after transplantation1, which is a curative therapy for numerous diseases including immunodeficiencies and leukaemias2. Although substantial effort has been applied to identifying HSC maintenance factors through the characterization of the in vivo bone-marrow HSC microenvironment or niche3-5, stable ex vivo HSC expansion has previously been unattainable6,7. Here we describe the development of a defined, albumin-free culture system that supports the long-term ex vivo expansion of functional mouse HSCs. We used a systematic optimization approach, and found that high levels of thrombopoietin synergize with low levels of stem-cell factor and fibronectin to sustain HSC self-renewal. Serum albumin has long been recognized as a major source of biological contaminants in HSC cultures8; we identify polyvinyl alcohol as a functionally superior replacement for serum albumin that is compatible with good manufacturing practice. These conditions afford between 236- and 899-fold expansions of functional HSCs over 1 month, although analysis of clonally derived cultures suggests that there is considerable heterogeneity in the self-renewal capacity of HSCs ex vivo. Using this system, HSC cultures that are derived from only 50 cells robustly engraft in recipient mice without the normal requirement for toxic pre-conditioning (for example, radiation), which may be relevant for HSC transplantation in humans. These findings therefore have important implications for both basic HSC research and clinical haematology.