MilliporeSigma
  • Antioxidant effect of glutathione on promoting 2-keto-l-gulonic acid production in vitamin C fermentation system.
Antioxidant effect of glutathione on promoting 2-keto-l-gulonic acid production in vitamin C fermentation system.
Journal of applied microbiology (2018-07-28)
M Huang, Y-H Zhang, S Yao, D Ma, X-D Yu, Q Zhang, S-X Lyu
ABSTRACT
Oxidative stress limited the growth of cells and 2-keto-l-gulonic acid (2-KGA) production in vitamin C (Vc) fermentation system. The study aims to investigate the antioxidant effect of glutathione on promoting 2-KGA in Vc fermentation system using Ketogulonicigenium vulgare 25B-1 and Bacillus endophyticus ST-1 as the co-culturing microbes. The activities of antioxidant-related enzymes and qPCR were used to study the antioxidant effect of glutathione addition in Vc fermentation system. The addition of GSH and GSH/GSSG increased 2-KGA production and decreased fermentation time, and the highest 2-KGA production increased by 40·63% and the lowest fermentation time shortened to 60 h when the addition of optimal concentration ratio of GSH/GSSG was 50 : 1. Moreover, the increased production of 2-KGA was accompanied by up-regulated the activities of total antioxidant capacity (T-AOC), total superoxide dismutase (T-SOD), catalase (CAT) and over-expressed oxidative stress-related genes sod, gst, gr, zwf, gp, which resulted in scavenging reactive oxygen species to reduce oxidative stress in Vc fermentation system. Glutathione showed a significant effect on increasing 2-KGA production and decreasing fermentation time in Vc fermentation system. GSH/GSSG could maintain a dynamic balance with two forms of glutathione and the optimal concentration ratio of GSH/GSSG was 50 : 1. Glutathione is proved to be effective to relieve oxidative stress. The promotion effects of GSSG and GSH on 2-KGA production could help to further explore the optimization of co-culture fermentation process for Vc industrial production.