MilliporeSigma
  • Establishment of serine protease htrA mutants in Helicobacter pylori is associated with secA mutations.
Establishment of serine protease htrA mutants in Helicobacter pylori is associated with secA mutations.
Scientific reports (2019-08-15)
Anna Zawilak-Pawlik, Urszula Zarzecka, Dorota Żyła-Uklejewicz, Jakub Lach, Dominik Strapagiel, Nicole Tegtmeyer, Manja Böhm, Steffen Backert, Joanna Skorko-Glonek
ABSTRACT
Helicobacter pylori plays an essential role in the pathogenesis of gastritis, peptic ulcer disease, and gastric cancer. The serine protease HtrA, an important secreted virulence factor, disrupts the gastric epithelium, which enables H. pylori to transmigrate across the epithelium and inject the oncogenic CagA protein into host cells. The function of periplasmic HtrA for the H. pylori cell is unknown, mainly due to unavailability of the htrA mutants. In fact, htrA has been described as an essential gene in this bacterium. We have screened 100 worldwide H. pylori isolates and show that only in the N6 strain it was possible to delete htrA or mutate the htrA gene to produce proteolytically inactive HtrA. We have sequenced the wild-type and mutant chromosomes and we found that inactivation of htrA is associated with mutations in SecA - a component of the Sec translocon apparatus used to translocate proteins from the cytoplasm into the periplasm. The cooperation of SecA and HtrA has been already suggested in Streptococcus pneumonia, in which these two proteins co-localize. Hence, our results pinpointing a potential functional relationship between HtrA and the Sec translocon in H. pylori possibly indicate for the more general mechanism responsible to maintain bacterial periplasmic homeostasis.