MilliporeSigma
  • ENEA, a peach and apricot IgE-binding protein cross-reacting with the latex major allergen Hev b 5.
ENEA, a peach and apricot IgE-binding protein cross-reacting with the latex major allergen Hev b 5.
Molecular immunology (2019-06-30)
Ivana Giangrieco, Teresa Ricciardi, Claudia Alessandri, Lucia Farina, Roberta Crescenzo, Lisa Tuppo, Michela Ciancamerla, Chiara Rafaiani, Maria Livia Bernardi, Anna Filomena Digilio, Beatrice Cobucci-Ponzano, Maurizio Tamburrini, Adriano Mari, Maria Antonietta Ciardiello
ABSTRACT
Peach and apricot can cause allergic reactions with symptoms ranging from mild to very severe, including anaphylaxis. Sometimes subjects allergic to fruits of the Prunus genus have been reported to be also allergic to rubber latex products. The objective of this study is the characterization of a newly identified peach and apricot protein showing similarities with the allergens Hev b 5 from rubber latex and Man e 5 from manioc. This protein has been named ENEA on the basis of the single letter amino acid code of the first four N-terminal residues of the isolated molecule. It has been found in very variable amounts in different peach cultivars and batches. ENEA was isolated from peach pulp extracts by chromatographic separations and identified by direct protein sequencing. At that time, the full length sequence was available only for the homologous protein of the taxonomically closely related apricot, which was produced as a recombinant molecule in Escherichia coli. The following availability of the full length sequence of peach ENEA revealed a very high identity (97%) with the apricot homolog. Similarly to Hev b 5 and to Man e 5, the structural characterization indicated that ENEA is an intrinsically disordered protein. The immunological properties, investigated by dot blotting, the ABA system and the FABER test, showed that ENEA is recognized by specific IgE of allergic patients. In a selected population of 31 patients reporting allergic reactions to peach fruit and/or IgE positive to Hev b 5, 28 and 27 subjects resulted co-sensitized to rENEA and Hev b 5 in the ABA and ISAC test, respectively. In a random population of 3305 suspected allergic patients, analyzed with the FABER test, 17 of them were sensitized to rENEA and 10 of them were also positive to Hev b 5. In addition, both the natural molecule from peach and the recombinant protein of apricot partially inhibited the IgE binding to Hev b 5. In conclusion, a new peach and apricot IgE-binding protein, cross-reacting with the major latex allergen Hev b 5, has been identified. Its variable concentration in the fruit might explain some occasionally occurring allergic reactions. The apricot molecule has recently been registered by the WHO/IUIS Allergen Nomenclature Sub-Committee with the allergen name Pru ar 5. The recombinant form of apricot ENEA, now available, will contribute to allergy diagnosis.