MilliporeSigma
  • The evolutionarily conserved indolergic receptors of the non-hematophagous elephant mosquito Toxorhynchites amboinensis.
The evolutionarily conserved indolergic receptors of the non-hematophagous elephant mosquito Toxorhynchites amboinensis.
Insect biochemistry and molecular biology (2019-04-21)
Amir Dekel, Esther Yakir, Jonathan D Bohbot
ABSTRACT
The conservation of the mosquito indolergic receptors across the Culicinae and Anophelinae mosquito lineages, which spans 200 million years of evolution, is a testament to the central role of indolic compounds in the biology of these insects. Indole and skatole have been associated with the detection of oviposition sites and animal hosts. To evaluate the potential ecological role of these two compounds, we have used a pharmacological approach to characterize homologs of the indolergic receptors Or2 and Or10 in the non-hematophagous elephant mosquito Toxorhynchites amboinensis. We provide evidence that both receptors are narrowly tuned to indole and skatole like their counterparts from hematophagous mosquitoes. These findings indicate that Toxorhynchites detects indole and skatole in an ecological context to be determined and underscore the importance of understanding the role of these compounds in mosquitoes.