MilliporeSigma
  • Ionic Liquid Composite Polybenzimidazol Membranes for High Temperature PEMFC Applications.
Ionic Liquid Composite Polybenzimidazol Membranes for High Temperature PEMFC Applications.
Polymers (2019-04-25)
Jorge Escorihuela, Abel García-Bernabé, Álvaro Montero, Óscar Sahuquillo, Enrique Giménez, Vicente Compañ
ABSTRACT
A series of proton exchange membranes based on polybenzimidazole (PBI) were prepared using the low cost ionic liquids (ILs) derived from 1-butyl-3-methylimidazolium (BMIM) bearing different anions as conductive fillers in the polymeric matrix with the aim of enhancing the proton conductivity of PBI membranes. The composite membranes prepared by casting method (containing 5 wt. % of IL) exhibited good thermal, dimensional, mechanical, and oxidative stability for fuel cell applications. The effects of anion, temperature on the proton conductivity of phosphoric acid-doped membranes were systematically investigated by electrochemical impedance spectroscopy. The PBI composite membranes containing 1-butyl-3-methylimidazolium-derived ionic liquids exhibited high proton conductivity of 0.098 S·cm-1 at 120 °C when tetrafluoroborate anion was present in the polymeric matrix. This conductivity enhancement might be attributed to the formed hydrogen-bond networks between the IL molecules and the phosphoric acid molecules distributed along the polymeric matrix.