MilliporeSigma
  • A long non-coding RNA TSLD8 inhibits hepatocellular carcinoma by stabilizing WWOX.
A long non-coding RNA TSLD8 inhibits hepatocellular carcinoma by stabilizing WWOX.
Biochemical and biophysical research communications (2019-06-25)
Fengliang Xu, Botian Wang, Mingxia Liu, Tao Liu, Ronghua Zhang
ABSTRACT
The hepatocellular carcinoma (HCC) is a common and highly aggressive malignancy especially in China. Accumulating data have shown a critical role of long non-coding RNAs (lncRNAs) during cancer progression. However, the function of lncRNA TSLD8 remains elusive. By lncRNA profiling, we identify a novel lncRNA termed TSLD8 in HCC. TSLD8 expression is significantly lowered in HCC tissues and cell lines. TSLD8 facilitates migration and viability in SMMC-7721 and HepG2 cells. Furthermore, TSLD8 can interact with WWOX and protect WWOX from proteasome-mediated degradation. Using PuPGEA-based nanocomplex for gene delivery, we found that co-delivery of TSLD8 and WWOX may exhibit synergistic and additive effects to inhibit HCC progression. PuPGEA-based nanocomplex delivery does not substantially alter the blood chemistries (e.g. alkaline phosphatase, blood urea nitrogen, aspartate aminotransferase, alanine aminotransferase) or initiate immune responses implying a safe strategy. Collectively, our current study has identified a novel tumor suppressive lncRNA TSLD8 which exerts its tumor suppressive function by stabilizing WWOX. Co-delivery of TSLD8 and WWOX via PuPGEA-based nanocomplexes might provide promising therapeutics for eradicating HCC.