MilliporeSigma
  • Axin-1 binds to Caveolin-1 to regulate the LPS-induced inflammatory response in AT-I cells.
Axin-1 binds to Caveolin-1 to regulate the LPS-induced inflammatory response in AT-I cells.
Biochemical and biophysical research communications (2019-04-08)
Yongjuan Zhang, Haihua Luo, Xuejun Lv, Junyan Liu, Xuxin Chen, Yuying Li, Aihua Liu, Yong Jiang
ABSTRACT
Caveolin-1 has been reported to play an important role in the pathogenesis of acute respiratory distress syndrome (ARDS). This study was designed to identify Caveolin-1-interacting proteins to reveal the molecular mechanisms of ARDS. Yeast two-hybrid screening was performed using Caveolin-1 as the bait, and Axin-1 was identified as a binding partner for Caveolin-1. Co-immunoprecipitation demonstrated that the binding domains were located in the N-terminal region (1-100 aa) of Caveolin-1 and the C-terminal region (710-797 aa) of Axin-1. Caveolin-1 gene knockout or Axin-1 knockdown significantly decreased the levels of TNF-α and IL-6 in the supernatants of alveolar type I (AT-I) epithelial cells treated with LPS. Disrupting the interaction between Caveolin-1 and Axin-1 using CRISPR/Cas9 technology led to a significant increase in TNF-α and IL-6 from AT-I cells, along with a significant reduction in β-catenin expression. In conclusion, Axin-1 functions as an adaptor of Caveolin-1 and affects the production of inflammatory cytokines in AT-I cells challenged with LPS via β-catenin-mediated negative regulation.