MilliporeSigma
  • A Spectroscopic Study of Tautomeric Equilibrium of Salicylideneaniline in ZSM-5 Zeolites.
A Spectroscopic Study of Tautomeric Equilibrium of Salicylideneaniline in ZSM-5 Zeolites.
Molecules (Basel, Switzerland) (2019-03-01)
Matthieu Hureau, Alain Moissette, Konstantin S Smirnov
ABSTRACT
Salicylideneaniline (SA) sorbed in cation-exchanged M-ZSM-5 (M = H⁺, Li⁺, Na⁺, K⁺, Rb⁺, Cs⁺ and Zn2+) zeolites was studied by spectroscopic techniques assisted by quantum-chemical calculations. The nature of extra-framework cations present in the zeolite void was found to affect the spectral signature of the sorbed SA molecule that points to the shift of tautomeric equilibrium between the enol and keto forms. Small size cations, such as H⁺ and Li⁺, stabilize a cis-keto SA tautomer along with a enol one in the zeolite structure. The calculations indicate that the sorbed cis-keto tautomer may have the dipole large enough to be considered as a zwitterion. New features appearing in the spectra with the increase of the cation size were attributed to the presence of trans-keto SA tautomer, which up to now has been observed only in time-resolved spectroscopic experiments. A strong interaction of the molecule with cations in Zn-ZSM-5 zeolite results in the chelation of enol SA with the divalent Zn2+ ions. The results of the study suggest that the tautomeric equilibrium of molecules belonging to the Schiff base family can be tuned by the confinement in the nanoporous materials via a choice of topology of zeolite framework and the nature of extra-framework cations.