MilliporeSigma
  • Translating chemometric analysis into physiological insights from in vivo confocal Raman spectroscopy of the human stratum corneum.
Translating chemometric analysis into physiological insights from in vivo confocal Raman spectroscopy of the human stratum corneum.
Biochimica et biophysica acta. Biomembranes (2018-11-23)
Stefania Perticaroli, Dawn J Yeomans, Fred C Wireko, John T Webber, Kathy M Werchowski, R Thomas Cambron, Paula J Ray
ABSTRACT
The superficial layer of the skin, the stratum corneum (SC), consists of corneocytes surrounded by lipid regions and acts as a protective barrier for the body against water loss, toxic agents and microorganisms. As most substances permeate the stratum corneum through the lipid regions, lipid organization is considered crucial for the skin barrier function. Here, we investigate the potential of in vivo confocal Raman spectroscopy to describe the composition and organization of the SC. Confocal Raman spectroscopy is finding increasing use in the characterization of skin in biomedical, pharmaceutical and cosmetic applications. In this work, we analyze the spectra using chemometric methods and obtain principal components that correspond to the primary skin constituents: protein (keratin), natural moisturizing factor (NMF), water and lipid contributions in both ordered (orthorhombic) and disordered structural organization. By identifying these important components of the SC, these results highlight the utility of this in vivo, non-invasive, and depth resolved tool at the forefront of skin research.