MilliporeSigma
  • Synthesis and Physicochemical Evaluation of Entecavir-Fatty Acid Conjugates in Reducing Food Effect on Intestinal Absorption.
Synthesis and Physicochemical Evaluation of Entecavir-Fatty Acid Conjugates in Reducing Food Effect on Intestinal Absorption.
Molecules (Basel, Switzerland) (2018-03-23)
Hyuck Jun Jung, Myoung Jin Ho, Sungwan Ahn, Young Taek Han, Myung Joo Kang
ABSTRACT
The oral bioavailability of entecavir (EV), an anti-viral agent commonly prescribed to treat hepatitis B infections, is drastically reduced under a post-prandial state. This is primarily due to its low permeability in the gastrointestinal tract. To reduce the food effect on the intestinal absorption of the nucleotide analogue, four lipidic prodrugs were synthesized via the esterification of the primary alcohol of EV with fatty acids (hexanoic acid, octanoic acid, decanoic acid, and dodecanoic acid). EV-3-dodecanoate (or EV-C12) exhibited high solubility in a fed state simulated intestinal fluid (78.8 μg/mL), with the acceptable calculated logP value (3.62) and the lowest hydrolysis rate (22.5% for 12 h in simulated gastric fluid, pH 1.2). Therefore, it was chosen as a candidate to improve intestinal absorption of EV, especially under a fed state condition. Physical characterization using scanning electron microscopy, a differential scanning calorimeter, and X-ray powder diffraction revealed that EV-C12 had a rectangular-shaped crystalline form, with a melting point of about 170 °C. In a release test in biorelevant media, such as fasted and fed state-simulated intestinal and/or gastric fluid, more than 90% of the prodrug was released within 2 h in all media tested. These data suggest that this lipidic prodrug might have the potential to alleviate the negative food effect on the intestinal absorption of EV with increased therapeutic efficacy and patient compliance.