MilliporeSigma
  • Metabolomics-Driven Exploration of the Chemical Drug Space to Predict Combination Antimicrobial Therapies.
Metabolomics-Driven Exploration of the Chemical Drug Space to Predict Combination Antimicrobial Therapies.
Molecular cell (2019-05-03)
Adrian I Campos, Mattia Zampieri
ABSTRACT
Alternative to the conventional search for single-target, single-compound treatments, combination therapies can open entirely new opportunities to fight antibiotic resistance. However, combinatorial complexity prohibits experimental testing of drug combinations on a large scale, and methods to rationally design combination therapies are lagging behind. Here, we developed a combined experimental-computational approach to predict drug-drug interactions using high-throughput metabolomics. The approach was tested on 1,279 pharmacologically diverse drugs applied to the gram-negative bacterium Escherichia coli. Combining our metabolic profiling of drug response with previously generated metabolic and chemogenomic profiles of 3,807 single-gene deletion strains revealed an unexpectedly large space of inhibited gene functions and enabled rational design of drug combinations. This approach is applicable to other therapeutic areas and can unveil unprecedented insights into drug tolerance, side effects, and repurposing. The compendium of drug-associated metabolome profiles is available at https://zampierigroup.shinyapps.io/EcoPrestMet, providing a valuable resource for the microbiological and pharmacological communities.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Xanthine Oxidase microbial, lyophilized powder, ≥7 units/mg solid
Pricing and availability is not currently available.